Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102500.1 | NP_001095970.1 | 4 - 5 | 983 | 92 | |
GenBank mRNA | BC142240.1 | - | 4 - 5 | 983 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |