Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103089.2 | NP_001096559.1 | 1 - 2 | 220 | 102 | |
GenBank mRNA | BC151606.1 | - | 1 - 2 | 221 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |