Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103089.2 | NP_001096559.1 | 6 - 7 | 866 | 73 | |
GenBank mRNA | BC151606.1 | - | 6 - 7 | 867 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |