Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103239.2 | NP_001096709.2 | 5 - 6 | 529 | 65 | |
GenBank mRNA | BC149102.1 | - | 5 - 6 | 529 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |