Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103255.2 | NP_001096725.1 | 3 - 4 | 1209 | 69 | |
GenBank mRNA | BC149387.1 | - | 3 - 4 | 1210 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |