Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103255.2 | NP_001096725.1 | 6 - 7 | 1800 | 94 | |
GenBank mRNA | BC149387.1 | - | 6 - 7 | 1801 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |