Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105255.1 | NP_001098725.1 | 14 - 15 | 1521 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |