Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105334.1 | NP_001098804.1 | 5 - 6 | 493 | 149 | |
GenBank mRNA | BC123726.1 | - | 5 - 6 | 493 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |