Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105336.3 | NP_001098806.2 | 4 - 5 | 368 | 96 | |
NM_001289991.1 | NP_001276920.1 | 3 - 4 | 243 | 96 | ||
NM_001289992.1 | NP_001276921.1 | 3 - 4 | 243 | 96 | ||
GenBank mRNA | BC102519.1 | - | 4 - 5 | 369 | 96 | |
DV802986.1 | - | 3 - 4 | 244 | 96 | ||
EH146865.1 | - | 4 - 5 | 369 | 96 | ||
FE015298.1 | - | 3 - 4 | 243 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |