Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105394.1 | NP_001098864.1 | 2 - 3 | 264 | 100 | |
GenBank mRNA | BC134544.1 | - | 2 - 3 | 264 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |