Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105617.2 | NP_001099087.2 | 2 | 1769 | 140 | |
GenBank mRNA | BC153291.1 | - | 1 | 2846 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |