Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109766.1 | NP_001103236.1 | 7 - 8 | 1441 | 117 | |
GenBank mRNA | BC123643.1 | - | 4 - 5 | 656 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |