Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109982.1 | NP_001103452.1 | 5 - 6 | 671 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |