Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110070.1 | NP_001103540.1 | 7 - 8 | 1016 | 63 | |
GenBank mRNA | BC113312.1 | - | 7 - 8 | 1016 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |