Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110788.1 | NP_001104258.1 | 4 - 5 | 1776 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |