Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113172.1 | NP_001106643.1 | 2 - 3 | 202 | 71 | |
GenBank mRNA | EU276061.1 | - | 2 - 3 | 202 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |