Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113538.1 | NP_001107010.1 | 3 - 4 | 486 | 76 | |
GenBank mRNA | BC140681.1 | - | 3 - 4 | 486 | 76 | |
EE252825.1 | - | 3 - 4 | 487 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |