Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114657.1 | NP_001108129.1 | 14 - 15 | 1441 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |