Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034759.2 | NP_001029931.2 | 2 - 3 | 540 | 80 | |
GenBank mRNA | BC102886.1 | - | 2 - 3 | 258 | 80 | |
BT021503.1 | - | 2 - 3 | 261 | 80 | ||
DV841387.1 | - | 2 - 3 | 541 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |