Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046290.2 | NP_001039755.2 | 8 - 9 | 1627 | 130 | |
GenBank mRNA | BC112556.1 | - | 8 - 9 | 1628 | 130 | |
DT857793.1 | - | 2 - 3 | 362 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |