Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 17 - 18 | 1511 | 103 | |
GenBank mRNA | DV907223.1 | - | 2 - 3 | 200 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |