Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 18 - 19 | 1563 | 69 | |
GenBank mRNA | DV907223.1 | - | 3 - 4 | 252 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |