Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046611.3 | NP_001040076.2 | 5 - 6 | 834 | 72 | |
NM_001314058.1 | NP_001300987.1 | 5 - 6 | 834 | 72 | ||
GenBank mRNA | BC105321.1 | - | 5 - 6 | 704 | 72 | |
DV882781.1 | - | 5 - 6 | 507 | 72 | ||
EV635520.1 | - | 5 - 6 | 822 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |