Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076902.2 | NP_001070370.2 | 2 - 3 | 284 | 115 | |
GenBank mRNA | EE341048.1 | - | 2 - 3 | 241 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |