Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076902.2 | NP_001070370.2 | 6 - 7 | 727 | 69 | |
GenBank mRNA | BC123601.1 | - | 5 - 6 | 668 | 69 | |
EE341048.1 | - | 6 - 7 | 684 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |