Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076902.2 | NP_001070370.2 | 7 - 8 | 842 | 95 | |
GenBank mRNA | BC123601.1 | - | 6 - 7 | 783 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |