Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C23H6orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077999.2 | NP_001071467.1 | 1 - 2 | 607 | 137 | |
GenBank mRNA | BC126615.1 | - | 1 - 2 | 219 | 137 | |
BC148131.1 | - | 1 - 2 | 1499 | 137 | ||
BT030562.1 | - | 1 - 2 | 607 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |