Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078150.2 | NP_001071618.1 | 1 - 2 | 192 | 72 | |
GenBank mRNA | BT030519.1 | - | 1 - 2 | 192 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Inventoried Probe spans exons |