Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098084.1 | NP_001091553.1 | 7 - 8 | 747 | 78 | |
GenBank mRNA | BC140592.1 | - | 7 - 8 | 747 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |