Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098100.1 | NP_001091569.1 | 1 - 2 | 234 | 60 | |
GenBank mRNA | BC140600.1 | - | 1 - 2 | 234 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |