Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098116.1 | NP_001091585.1 | 4 - 5 | 1900 | 95 | |
GenBank mRNA | BC140469.1 | - | 4 - 5 | 1900 | 95 | |
BC149610.1 | - | 4 - 5 | 1542 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |