Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098123.1 | NP_001091592.1 | 3 - 4 | 1631 | 109 | |
GenBank mRNA | BC140519.1 | - | 3 - 4 | 1631 | 109 | |
BC149076.1 | - | 1 - 2 | 1155 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |