Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098137.1 | NP_001091606.1 | 3 - 4 | 332 | 148 | |
GenBank mRNA | BC140690.1 | - | 3 - 4 | 332 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |