Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099071.2 | NP_001092541.1 | 4 - 5 | 870 | 84 | |
GenBank mRNA | BC142398.1 | - | 4 - 5 | 871 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |