Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099126.2 | NP_001092596.1 | 1 - 2 | 294 | 75 | |
GenBank mRNA | BC146138.1 | - | 1 - 2 | 295 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |