Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099380.2 | NP_001092850.1 | 8 - 9 | 1185 | 85 | |
GenBank mRNA | BC140686.1 | - | 8 - 9 | 1186 | 85 | |
BT029788.1 | - | 5 - 6 | 692 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |