Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099381.1 | NP_001092851.1 | 4 - 5 | 662 | 90 | |
GenBank mRNA | BC146132.1 | - | 4 - 5 | 662 | 90 | |
BT025341.1 | - | 4 - 5 | 660 | 90 | ||
BT025347.1 | - | 4 - 5 | 633 | 90 | ||
BT025488.1 | - | 4 - 5 | 617 | 90 | ||
BT026290.1 | - | 4 - 5 | 611 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |