Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099709.2 | NP_001093179.1 | 3 - 4 | 740 | 70 | |
GenBank mRNA | BC142341.1 | - | 3 - 4 | 741 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |