Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101127.1 | NP_001094597.1 | 7 - 8 | 908 | 73 | |
GenBank mRNA | BC147929.1 | - | 7 - 8 | 908 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |