Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101127.1 | NP_001094597.1 | 9 - 10 | 1102 | 99 | |
GenBank mRNA | BC147929.1 | - | 9 - 10 | 1102 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |