Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101136.1 | NP_001094606.1 | 4 - 5 | 401 | 82 | |
GenBank mRNA | BC148002.1 | - | 4 - 5 | 401 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |