Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101151.1 | NP_001094621.1 | 6 - 7 | 697 | 117 | |
GenBank mRNA | BC147994.1 | - | 6 - 7 | 697 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |