Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101168.1 | NP_001094638.1 | 3 - 4 | 309 | 105 | |
GenBank mRNA | BC148140.1 | - | 3 - 4 | 309 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |