Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101168.1 | NP_001094638.1 | 7 - 8 | 688 | 71 | |
GenBank mRNA | BC133555.1 | - | 4 - 5 | 381 | 71 | |
BC148140.1 | - | 7 - 8 | 688 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |