Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101168.1 | NP_001094638.1 | 8 - 9 | 817 | 69 | |
GenBank mRNA | BC133555.1 | - | 5 - 6 | 510 | 69 | |
BC148140.1 | - | 8 - 9 | 817 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |