Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101172.1 | NP_001094642.1 | 3 - 4 | 651 | 87 | |
GenBank mRNA | BC149365.1 | - | 3 - 4 | 651 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |