Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101192.2 | NP_001094662.1 | 3 - 4 | 423 | 56 | |
GenBank mRNA | BC151528.1 | - | 3 - 4 | 424 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |