Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101237.1 | NP_001094707.1 | 5 - 6 | 363 | 143 | |
GenBank mRNA | BC151658.1 | - | 5 - 6 | 363 | 143 | |
EH141890.1 | - | 5 - 6 | 363 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |