Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101241.1 | NP_001094711.1 | 5 - 6 | 763 | 70 | |
GenBank mRNA | BC149224.1 | - | 5 - 6 | 763 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |