Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101247.1 | NP_001094717.1 | 5 - 6 | 336 | 142 | |
GenBank mRNA | BC149943.1 | - | 5 - 6 | 336 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |